Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPT1B
Entrez Gene ID
1375
Gene Name
carnitine palmitoyltransferase 1B
Gene Aliases
CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1
Chromosome Location
Chr.22: 50568861 - 50578667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004377.3 NP_004368.1 19 - 19 2613 133
NM_152245.2 NP_689451.1 19 - 19 2652 133

View Full Product Details