Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEAL3
Entrez Gene ID
85012
Gene Name
transcription elongation factor A like 3
Gene Aliases
WEX8
Chromosome Location
Chr.X: 103607906 - 103609927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006933.1 NP_001006934.1 1 - 2 168 107
GenBank mRNA AF163263.1 - 1 - 2 105 107

View Full Product Details