Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NPY6R
- Entrez Gene ID
-
4888
- Gene Name
- neuropeptide Y receptor Y6 (pseudogene)
- Gene Aliases
-
NPY1RL, NPY6RP, PP2, Y2B
- Chromosome Location
-
Chr.5: 137801193 - 137810751 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002713.3 |
|
2 - 2 |
1856 |
136 |
GenBank mRNA |
AK297855.1 |
- |
1 - 1 |
1364 |
136 |
D86519.1 |
- |
1 - 1 |
1847 |
136 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
heterogeneous nuclear ribonucleoprotein A0 |
HNRNPA0 |
HNRPA0 |
-53.7kb
|
Anti-sense |
Chr. - 137751384 - 137754350 on Build GRCh38 |
Downstream |
myotilin |
MYOT |
LGMD1, LGMD1A, MFM3, TTID, TTOD |
57.1kb
|
Sense |
Chr. - 137867282 - 137887851 on Build GRCh38 |
View Full Product Details