Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD4A
- Entrez Gene ID
-
26773
- Gene Name
- small nucleolar RNA, C/D box 4A
- Gene Aliases
-
RNU101A, Z17A, mgh18S-121
- Chromosome Location
-
Chr.17: 28722582 - 28722653 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_000010.1 |
|
1 - 1 |
36 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM979034.1 |
- |
3 |
|
|
Chr. 17 - 28722445 - 28723940 [-] on Build GRCh38 |
69 |
GenBankmRNA |
BQ300200.1 |
- |
1 |
136 |
|
Chr. 17 - 28722484 - 28722832 [+] on Build GRCh38 |
69 |
GenBankmRNA |
BQ575751.1 |
- |
3 |
|
|
Chr. 17 - 28721890 - 28723940 [-] on Build GRCh38 |
69 |
GenBankmRNA |
AL569524.3 |
- |
1 |
|
|
Chr. 17 - 28721450 - 28722880 [-] on Build GRCh38 |
69 |
GenBankmRNA |
CB242853.1 |
- |
3 |
|
|
Chr. 17 - 28721939 - 28723940 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 42B |
SNORD42B |
RNU42B, U42B |
-2.0kb
|
Sense |
Chr. - 28720550 - 28720616 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 42A |
SNORD42A |
RNU42A, U42, U42A |
777.0
|
Sense |
Chr. - 28723430 - 28723491 on Build GRCh38 |
View Full Product Details