Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HAS2-AS1
- Entrez Gene ID
-
594842
- Gene Name
- HAS2 antisense RNA 1
- Gene Aliases
-
HAS2-AS, HAS2AS, HASNT, NCRNA00077
- Chromosome Location
-
Chr.8: 121639346 - 121645325 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002835.2 |
|
3 - 4 |
1229 |
149 |
GenBank mRNA |
AY941178.1 |
- |
3 - 4 |
1229 |
149 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CA336013.1 |
- |
2 - 3 |
168 |
|
Chr. 8 - 121641436 - 121644693 [+] on Build GRCh38 |
149 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
hyaluronan synthase 2 |
HAS2 |
|
-1.2kb
|
Anti-sense |
Chr. - 121613031 - 121641390 on Build GRCh38 |
Downstream |
uncharacterized LOC105375734 |
LOC105375734 |
|
768.4kb
|
Anti-sense |
Chr. - 122414327 - 122568644 on Build GRCh38 |
View Full Product Details