Product Details

Assay Reference Genome
Location

Chr.1:12169040 on build GRCh38
Cytoband
1p36.22
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7133
Gene Symbol
TNFRSF1B
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
Location
Chr.1:12166948-12209222 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TNFRSF1B NM_001066.2 NP_001057.1
XM_011542060.2 XP_011540362.1
XM_011542063.2 XP_011540365.1
XM_017002211.1 XP_016857700.1
XM_017002214.1 XP_016857703.1
XM_017002215.1 XP_016857704.1
AY148473.1 AAN72434.1
BC011844.2 AAH11844.1
BC042167.1 AAH42167.1
BC052977.1 AAH52977.1
BT019927.1 AAV38730.1
EU826615.1
M32315.1 AAA59929.1
M55994.1 AAA36755.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv471622 Chr1:12155207 - 12371588 on Build GRCh38 Gain+Loss VPS13D TNFRSF1B MIR4632 MIR7846
nsv508891 Chr1:12097920 - 12235045 on Build GRCh38 Insertion VPS13D TNFRSF8 TNFRSF1B MIR4632 MIR7846
esv3584475 Chr1:12134394 - 12217607 on Build GRCh38 Gain TNFRSF8 TNFRSF1B MIR4632 MIR7846
nsv527024 Chr1:12164182 - 12207208 on Build GRCh38 Loss TNFRSF1B MIR4632 MIR7846
nsv469830 Chr1:12039669 - 12199858 on Build GRCh38 Loss TNFRSF8 TNFRSF1B MIR4632 MIR7846

View Full Product Details