Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
TNFRSF1B | NM_001066.2 | NP_001057.1 | ||
XM_011542060.2 | XP_011540362.1 | |||
XM_011542063.2 | XP_011540365.1 | |||
XM_017002211.1 | XP_016857700.1 | |||
XM_017002212.1 | XP_016857701.1 | |||
XM_017002213.1 | XP_016857702.1 | |||
XM_017002214.1 | XP_016857703.1 | |||
XM_017002215.1 | XP_016857704.1 | |||
AY148473.1 | AAN72434.1 | |||
BC011844.2 | AAH11844.1 | |||
BC042167.1 | AAH42167.1 | |||
BC052977.1 | AAH52977.1 | |||
BT019927.1 | AAV38730.1 | |||
EU826615.1 | ||||
M32315.1 | AAA59929.1 | |||
M55994.1 | AAA36755.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471622 | Chr1:12155207 - 12371588 on Build GRCh38 | Gain+Loss | MIR4632 VPS13D MIR7846 TNFRSF1B |
nsv508891 | Chr1:12097920 - 12235045 on Build GRCh38 | Insertion | MIR4632 VPS13D MIR7846 TNFRSF8 TNFRSF1B |
esv3584475 | Chr1:12134394 - 12217607 on Build GRCh38 | Gain | MIR4632 MIR7846 TNFRSF8 TNFRSF1B |
nsv527024 | Chr1:12164182 - 12207208 on Build GRCh38 | Loss | MIR4632 MIR7846 TNFRSF1B |
nsv469830 | Chr1:12039669 - 12199858 on Build GRCh38 | Loss | MIR4632 MIR7846 TNFRSF8 TNFRSF1B |