The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005353.2 | NP_001005353.1 | 3 - 4 | 621 | 134 |
NM_013410.3 | NP_037542.1 | 2 - 3 | 504 | 134 | |
NM_203464.2 | NP_982289.1 | 3 - 4 | 510 | 134 | |
XM_017000612.1 | XP_016856101.1 | 2 - 3 | 256 | 134 | |
XM_017000613.1 | XP_016856102.1 | 2 - 3 | 442 | 134 | |
GenBank mRNA | AK025926.1 | - | 2 - 3 | 468 | 134 |
AK309057.1 | - | 2 - 3 | 212 | 134 | |
AK313611.1 | - | 2 - 3 | 468 | 134 | |
BC016180.1 | - | 2 - 3 | 404 | 134 | |
BC040224.1 | - | 2 - 3 | 380 | 134 | |
BC066944.1 | - | 3 - 4 | 621 | 134 | |
BC136886.1 | - | 2 - 3 | 345 | 134 | |
BC136887.1 | - | 1 - 1 | 345 | 134 | |
BC146653.1 | - | 3 - 4 | 495 | 134 | |
BC148270.1 | - | 2 - 3 | 285 | 134 | |
BF205061.1 | - | 1 - 1 | 467 | 133 | |
BU164554.1 | - | 3 - 4 | 467 | 134 | |
CR456830.1 | - | 2 - 3 | 263 | 134 |