The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002713.3 | 1 - 2 | 261 | 113 | |
GenBank mRNA | AK299427.1 | - | 1 - 2 | 128 | 113 |
DC357138.1 | - | 1 - 2 | 261 | 113 |