Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC646214
- Entrez Gene ID
-
646214
- Gene Name
- p21 protein (Cdc42/Rac)-activated kinase 2 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 21289604 - 21297829 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027053.2 |
|
1 - 1 |
956 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1193 |
LINC01193 |
CT60 |
-296.3kb
|
Anti-sense |
Chr. - 20940438 - 20993304 on Build GRCh38 |
Downstream |
coxsackie virus and adenovirus receptor pseudogene 2 |
CXADRP2 |
|
73.7kb
|
Sense |
Chr. - 21371493 - 21373968 on Build GRCh38 |
View Full Product Details