Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP424608.1 |
- |
1 |
403 |
|
Chr. 4 - 121031569 - 121032072 [-] on Build GRCh38 |
102 |
GenBankmRNA |
BM676840.1 |
- |
1 |
|
|
Chr. 4 - 121031418 - 121032054 [+] on Build GRCh38 |
102 |
GenBankmRNA |
BX101026.1 |
- |
1 |
190 |
|
Chr. 4 - 121031299 - 121031863 [-] on Build GRCh38 |
102 |
GenBankmRNA |
BM673795.1 |
- |
1 |
|
|
Chr. 4 - 121031425 - 121032094 [+] on Build GRCh38 |
102 |
GenBankmRNA |
AL049990.1 |
- |
1 |
1447 |
|
Chr. 4 - 121031418 - 121033117 [-] on Build GRCh38 |
102 |
GenBankmRNA |
DB326607.1 |
- |
1 |
|
|
Chr. 4 - 121031420 - 121031952 [+] on Build GRCh38 |
102 |
GenBankmRNA |
AW614170.1 |
- |
1 |
|
|
Chr. 4 - 121031419 - 121031853 [+] on Build GRCh38 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
PR domain 5 |
PRDM5 |
BCS2, PFM2 |
-108.6kb
|
Sense |
Chr. - 120684907 - 120922866 on Build GRCh38 |
Downstream |
neuron derived neurotrophic factor |
NDNF |
C4orf31, NORD |
2.5kb
|
Sense |
Chr. - 121035627 - 121072518 on Build GRCh38 |
View Full Product Details