Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ712619.1 - 1 515 Chr. 14 - 74082662 - 74083582 [-] on Build GRCh38 102
GenBankmRNA AK123178.1 - 1 847 Chr. 14 - 74082420 - 74083925 [-] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream basal body orientation factor 1 BBOF1 C14orf45, CCDC176 -4.1kb Anti-sense Chr. - 74019350 - 74078351 on Build GRCh38
Downstream lin-52 DREAM MuvB core complex component LIN52 C14orf46, c14_5549 911.0 Anti-sense Chr. - 74084831 - 74201255 on Build GRCh38

View Full Product Details