Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DHRS7B
- Entrez Gene ID
-
25979
- Gene Name
- dehydrogenase/reductase 7B
- Gene Aliases
-
CGI-93, SDR32C1
- Chromosome Location
-
Chr.17: 21123369 - 21191972 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124055.1 |
- |
1 - 1 |
845 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1563 |
LINC01563 |
HP08942 |
-36.4kb
|
Sense |
Chr. - 21075556 - 21090613 on Build GRCh38 |
Downstream |
transmembrane protein 11 |
TMEM11 |
C17orf35, PM1, PMI |
69.1kb
|
Anti-sense |
Chr. - 21197950 - 21214595 on Build GRCh38 |
View Full Product Details