Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK127825.1 - 1 945 Chr. 14 - 76494324 - 76495830 [-] on Build GRCh38 82


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream G-patch domain containing 2 like GPATCH2L C14orf118 -279.9kb Anti-sense Chr. - 76151885 - 76214473 on Build GRCh38
Downstream vasohibin 1 VASH1 KIAA1036 266.1kb Anti-sense Chr. - 76761457 - 76783020 on Build GRCh38

View Full Product Details