Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK128178.1 - 1 4352 Chr. 8 - 39479813 - 39484748 [-] on Build GRCh38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ADAM metallopeptidase domain 5 (pseudogene) ADAM5 ADAM5P, TMDCII -62.6kb Anti-sense Chr. - 39314663 - 39417226 on Build GRCh38
Downstream ADAM metallopeptidase domain 3A-like LOC100130964 78.4kb Anti-sense Chr. - 39563112 - 39580134 on Build GRCh38

View Full Product Details