Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX640993.1 - 1 1446 Chr. 17 - 22420022 - 22424731 [+] on Build GRCh38 109


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized FLJ36000 FLJ36000 -6.3kb Sense Chr. - 22406019 - 22413741 on Build GRCh38
Downstream MT-RNR2-like 1 MTRNR2L1 HN1 98.4kb Sense Chr. - 22523111 - 22524665 on Build GRCh38

View Full Product Details