Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
92129
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 106900063 - 106903331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171706.1 NP_001165177.1 1 - 2 196 85
GenBank mRNA BC127250.1 - 1 - 2 196 85
BX106639.1 - 1 - 2 232 85

View Full Product Details