The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152517.2 | NP_689730.2 | 1 - 1 | 2248 | 100 |
GenBank mRNA | AK296828.1 | - | 2 - 2 | 2082 | 100 |
AW007843.1 | - | 1 - 1 | 100 |