The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006607.2 | NP_001006608.2 | 7 - 8 | 3059 | 95 |
XM_011524841.2 | XP_011523143.1 | 6 - 7 | 3040 | 95 | |
XM_011524842.2 | XP_011523144.1 | 7 - 8 | 3112 | 95 | |
XM_011524844.2 | XP_011523146.1 | 5 - 6 | 2959 | 95 | |
XM_011524846.2 | XP_011523148.1 | 4 - 5 | 2887 | 95 | |
GenBank mRNA | AK126994.1 | - | 7 - 8 | 1741 | 95 |
AY386262.1 | - | 7 - 8 | 1480 | 95 | |
BC144427.1 | - | 7 - 8 | 722 | 95 | |
BC144428.1 | - | 7 - 8 | 722 | 95 | |
JQ790563.1 | - | 7 - 8 | 616 | 95 | |
JQ790564.1 | - | 7 - 8 | 616 | 95 | |
JQ790568.1 | - | 7 - 8 | 616 | 95 | |
JQ790570.1 | - | 7 - 8 | 616 | 95 | |
JQ790571.1 | - | 7 - 8 | 616 | 95 | |
JQ790576.1 | - | 7 - 8 | 616 | 95 | |
JQ790591.1 | - | 4 - 5 | 391 | 95 | |
JQ790595.1 | - | 4 - 5 | 391 | 95 | |
JQ790598.1 | - | 6 - 7 | 544 | 95 | |
JQ790606.1 | - | 7 - 8 | 616 | 95 | |
JQ790617.1 | - | 7 - 8 | 616 | 95 | |
JQ790675.1 | - | 4 - 5 | 391 | 95 | |
JQ790677.1 | - | 4 - 5 | 391 | 95 | |
JQ790700.1 | - | 7 - 8 | 616 | 95 | |
JQ790723.1 | - | 4 - 5 | 391 | 95 | |
JQ790739.1 | - | 7 - 8 | 618 | 95 | |
JQ790740.1 | - | 7 - 8 | 618 | 95 | |
JQ790748.1 | - | 7 - 8 | 619 | 95 | |
JQ790750.1 | - | 7 - 8 | 618 | 95 | |
JQ790764.1 | - | 7 - 8 | 618 | 95 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014834.4 | NP_055649.4 | 7 - 8 | 3059 | 95 |
XM_005257879.4 | XP_005257936.1 | 7 - 8 | 3317 | 95 | |
XM_005257882.4 | XP_005257939.1 | 8 - 9 | 3503 | 95 | |
XM_011525539.2 | XP_011523841.1 | 5 - 6 | 3164 | 95 | |
XM_017025472.1 | XP_016880961.1 | 9 - 10 | 2916 | 95 | |
XM_017025473.1 | XP_016880962.1 | 7 - 8 | 563 | 95 | |
GenBank mRNA | BC040501.1 | - | 7 - 8 | 3059 | 95 |
CR749232.1 | - | 7 - 8 | 3060 | 95 | |
JQ790562.1 | - | 7 - 8 | 616 | 95 | |
JQ790566.1 | - | 7 - 8 | 616 | 95 | |
JQ790572.1 | - | 7 - 8 | 616 | 95 | |
JQ790577.1 | - | 7 - 8 | 616 | 95 | |
JQ790578.1 | - | 7 - 8 | 616 | 95 | |
JQ790579.1 | - | 7 - 8 | 616 | 95 | |
JQ790580.1 | - | 7 - 8 | 616 | 95 | |
JQ790581.1 | - | 7 - 8 | 616 | 95 | |
JQ790593.1 | - | 4 - 5 | 391 | 95 | |
JQ790594.1 | - | 4 - 5 | 391 | 95 | |
JQ790596.1 | - | 4 - 5 | 391 | 95 | |
JQ790688.1 | - | 7 - 8 | 616 | 95 | |
JQ790690.1 | - | 7 - 8 | 616 | 95 | |
JQ790691.1 | - | 7 - 8 | 616 | 95 | |
JQ790693.1 | - | 7 - 8 | 616 | 95 | |
JQ790694.1 | - | 7 - 8 | 616 | 95 | |
JQ790724.1 | - | 4 - 5 | 391 | 95 | |
JQ790725.1 | - | 4 - 5 | 391 | 95 | |
JQ790726.1 | - | 7 - 8 | 618 | 95 | |
JQ790727.1 | - | 7 - 8 | 618 | 95 | |
JQ790729.1 | - | 7 - 8 | 618 | 95 | |
JQ790730.1 | - | 7 - 8 | 618 | 95 | |
JQ790732.1 | - | 4 - 5 | 393 | 95 | |
JQ790738.1 | - | 7 - 8 | 618 | 95 | |
JQ790743.1 | - | 7 - 8 | 618 | 95 | |
JQ790744.1 | - | 7 - 8 | 617 | 95 | |
JQ790749.1 | - | 7 - 8 | 618 | 95 | |
JQ790752.1 | - | 7 - 8 | 618 | 95 | |
JQ790755.1 | - | 7 - 8 | 618 | 95 | |
JQ790756.1 | - | 7 - 8 | 618 | 95 | |
JQ790758.1 | - | 7 - 8 | 618 | 95 | |
JQ790763.1 | - | 7 - 8 | 619 | 95 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | JQ790698.1 | - | 7 - 8 | 616 | 95 |