Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBC1D3B
- Entrez Gene ID
-
414059
- Gene Name
- TBC1 domain family member 3B
- Gene Aliases
-
PRC17, TBC1D3I, TBC1D3L
- Chromosome Location
-
Chr.17: 36165681 - 36176624 on Build GRCh38
- Gene Symbol
- TBC1D3F
- Entrez Gene ID
-
84218
- Gene Name
- TBC1 domain family member 3F
- Gene Aliases
-
TBC1D3
- Chromosome Location
-
Chr.17: 36428612 - 36439523 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK225985.1 |
- |
7 - 8 |
602 |
85 |
- Gene Symbol
- TBC1D3G
- Entrez Gene ID
-
101060321
- Gene Name
- TBC1 domain family member 3G
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36322242 - 36334759 on Build GRCh38
- Gene Symbol
- TBC1D3H
- Entrez Gene ID
-
729877
- Gene Name
- TBC1 domain family member 3H
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36377475 - 36388448 on Build GRCh38
View Full Product Details