The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007038.4 | NP_008969.2 | 8 - 8 | 3418 | 73 |
XM_011529448.2 | XP_011527750.1 | 7 - 7 | 3250 | 73 | |
GenBank mRNA | AF141293.1 | - | 6 - 6 | 1453 | 73 |
AF142099.1 | - | 8 - 8 | 2811 | 73 | |
BC093775.1 | - | 8 - 8 | 2773 | 73 | |
BC093777.1 | - | 8 - 8 | 2773 | 73 |