Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSX8
Entrez Gene ID
280659
Gene Name
SSX family member 8
Gene Aliases
-
Chromosome Location
Chr.X: 52622935 - 52633948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027250.2 1 - 2 267 120
GenBank mRNA AK302291.1 - 1 - 2 268 120
DC399111.1 - 1 - 2 268 120


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream X antigen family member 1E XAGE1E CT12.1, CT12.1C, CT12.1D, CT12.1E, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1C, XAGE1D -105.7kb Anti-sense Chr. - 52512074 - 52517221 on Build GRCh38
Downstream SSX family member 7 SSX7 19.0kb Anti-sense Chr. - 52644061 - 52654900 on Build GRCh38

View Full Product Details