Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTAN1
Entrez Gene ID
123803
Gene Name
N-terminal asparagine amidase
Gene Aliases
PNAA, PNAD
Chromosome Location
Chr.16: 15037853 - 15056079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173474.3 NP_775745.1 1 - 2 186 98
GenBank mRNA BC017336.2 - 1 - 2 83 98
HY036316.1 - 1 - 2 187 98

View Full Product Details