Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
Chromosome Location
Chr.2: 143945787 - 144333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006636.3 NP_001006637.1 4 - 5 457 75
NM_001164629.3 NP_001158101.1 4 - 5 442 75
NM_001284233.1 NP_001271162.1 2 - 3 266 75
NM_001284235.1 NP_001271164.1 4 - 5 424 75
NM_024659.4 NP_078935.2 4 - 5 424 75
XM_005263774.2 XP_005263831.1 4 - 5 394 75
XM_005263782.2 XP_005263839.1 4 - 5 410 75
XM_005263784.2 XP_005263841.1 2 - 3 269 75
XM_011511843.2 XP_011510145.1 4 - 5 443 75
XM_011511855.2 XP_011510157.1 2 - 3 387 75
XM_017004917.1 XP_016860406.1 8 - 9 1065 75
XM_017004918.1 XP_016860407.1 6 - 7 657 75
XM_017004919.1 XP_016860408.1 4 - 5 543 75
XM_017004920.1 XP_016860409.1 6 - 7 3699 75
XM_017004921.1 XP_016860410.1 5 - 6 492 75
XM_017004922.1 XP_016860411.1 5 - 6 669 75
XM_017004923.1 XP_016860412.1 4 - 5 457 75
XM_017004926.1 XP_016860415.1 2 - 3 393 75
XM_017004927.1 XP_016860416.1 4 - 5 424 75
XM_017004928.1 XP_016860417.1 6 - 7 657 75
XM_017004929.1 XP_016860418.1 4 - 5 394 75
XM_017004930.1 XP_016860419.1 4 - 5 1045 75
XM_017004931.1 XP_016860420.1 4 - 5 458 75
XM_017004932.1 XP_016860421.1 4 - 5 428 75
XM_017004933.1 XP_016860422.1 4 - 5 410 75
XM_017004934.1 XP_016860423.1 5 - 6 3617 75
XM_017004935.1 XP_016860424.1 6 - 7 1487 75
XM_017004936.1 XP_016860425.1 5 - 6 664 75
XM_017004937.1 XP_016860426.1 2 - 3 398 75
XM_017004938.1 XP_016860427.1 2 - 3 396 75
XM_017004939.1 XP_016860428.1 4 - 5 446 75
XM_017004940.1 XP_016860429.1 2 - 3 394 75
XM_017004941.1 XP_016860430.1 2 - 3 393 75
XM_017004942.1 XP_016860431.1 4 - 5 440 75
XM_017004943.1 XP_016860432.1 4 - 5 1106 75
XM_017004944.1 XP_016860433.1 4 - 5 439 75
XM_017004945.1 XP_016860434.1 6 - 7 657 75
XM_017004946.1 XP_016860435.1 4 - 5 394 75
XM_017004947.1 XP_016860436.1 4 - 5 1077 75
XM_017004948.1 XP_016860437.1 5 - 6 3617 75
XM_017004949.1 XP_016860438.1 2 - 3 392 75
XM_017004950.1 XP_016860439.1 4 - 5 439 75
XM_017004951.1 XP_016860440.1 4 - 5 394 75
XM_017004952.1 XP_016860441.1 4 - 5 838 75
XM_017004953.1 XP_016860442.1 4 - 5 410 75
XM_017004954.1 XP_016860443.1 5 - 6 498 75
XM_017004955.1 XP_016860444.1 7 - 8 846 75
XM_017004956.1 XP_016860445.1 2 - 3 394 75
XM_017004957.1 XP_016860446.1 4 - 5 440 75
GenBank mRNA AB201269.1 - 1 - 2 178 75
AB597184.1 - 1 - 2 178 75
AK091684.1 - 2 - 3 270 75
AK291357.1 - 4 - 5 372 75
AY281366.1 - 4 - 5 457 75
BC017741.2 - 5 - 6 464 75
BC061699.1 - 4 - 5 364 75
DB458955.1 - 4 - 5 424 75

View Full Product Details