The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_108036.1 | 1 - 2 | 453 | 85 | |
GenBank mRNA | BX093559.1 | - | 1 - 2 | 384 | 85 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | cilia and flagella associated protein 58 | CFAP58 | C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 | Anti-sense | Chr. - 104339124 - 104455090 on Build GRCh38 | |
Downstream | uncharacterized LOC101927523 | LOC101927523 | 121.4kb | Sense | Chr. - 104474939 - 104480275 on Build GRCh38 |