The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301226.1 | NP_001288155.1 | 4 - 5 | 730 | 68 |
NM_001301227.1 | NP_001288156.1 | 4 - 5 | 730 | 68 | |
NM_003289.3 | NP_003280.2 | 4 - 5 | 730 | 68 | |
NM_213674.1 | NP_998839.1 | 4 - 5 | 730 | 68 | |
XM_017015087.1 | XP_016870576.1 | 4 - 5 | 602 | 68 | |
XM_017015088.1 | XP_016870577.1 | 4 - 5 | 602 | 68 | |
XM_017015089.1 | XP_016870578.1 | 4 - 5 | 602 | 68 | |
XM_017015090.1 | XP_016870579.1 | 4 - 5 | 602 | 68 | |
XM_017015091.1 | XP_016870580.1 | 4 - 5 | 602 | 68 | |
XM_017015092.1 | XP_016870581.1 | 4 - 5 | 602 | 68 | |
XM_017015093.1 | XP_016870582.1 | 4 - 5 | 602 | 68 | |
GenBank mRNA | AK130045.1 | - | 4 - 5 | 318 | 68 |
AK223258.1 | - | 4 - 5 | 599 | 68 | |
AK294522.1 | - | 4 - 5 | 599 | 68 | |
AK300249.1 | - | 3 - 4 | 694 | 68 | |
AK304803.1 | - | 4 - 5 | 599 | 68 | |
BC011776.2 | - | 4 - 5 | 550 | 68 | |
BE294047.1 | - | 3 - 4 | 291 | 68 | |
EU106177.1 | - | 4 - 5 | 491 | 68 | |
FJ224304.1 | - | 4 - 5 | 730 | 68 | |
M12125.1 | - | 4 - 5 | 609 | 68 | |
M74817.1 | - | 4 - 5 | 580 | 68 | |
M75165.1 | - | 4 - 5 | 580 | 68 | |
X06825.1 | - | 4 - 5 | 597 | 68 |