Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GTDC1
- Entrez Gene ID
-
79712
- Gene Name
- glycosyltransferase like domain containing 1
- Gene Aliases
-
Hmat-Xa, mat-Xa
- Chromosome Location
-
Chr.2: 143945787 - 144333092 on Build GRCh38
View Full Product Details