The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000769.2 | NP_000760.1 | 5 - 6 | 842 | 106 |
GenBank mRNA | M61854.1 | - | 5 - 6 | 822 | 106 |