Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101927948
- Entrez Gene ID
-
101927948
- Gene Name
- uncharacterized LOC101927948
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 78088730 - 78127806 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110287.1 |
|
3 - 4 |
354 |
111 |
GenBank mRNA |
BC042517.1 |
- |
3 - 4 |
354 |
111 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG723219.1 |
- |
3 - 4 |
358 |
|
Chr. 2 - 78088730 - 78112132 [+] on Build GRCh38 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927967 |
LOC101927967 |
|
|
Anti-sense |
Chr. - 77743696 - 78290731 on Build GRCh38 |
Downstream |
uncharacterized LOC105374820 |
LOC105374820 |
|
434.9kb
|
Sense |
Chr. - 78545531 - 78566088 on Build GRCh38 |
View Full Product Details