The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120622.1 | 1 - 2 | 203 | 84 | |
GenBank mRNA | BM794484.1 | - | 1 - 2 | 203 | 84 |
BX094877.1 | - | 1 - 2 | 180 | 84 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | glutathione S-transferase omega 2 | GSTO2 | GSTO 2-2, bA127L20.1 | -18.4kb | Sense | Chr. - 104267626 - 104304948 on Build GRCh38 |
Downstream | cilia and flagella associated protein 58 | CFAP58 | C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 | 14.5kb | Sense | Chr. - 104339124 - 104455090 on Build GRCh38 |