Product Details

Assay Reference Genome
Location

Chr.16:3590621 on build GRCh38
Cytoband
16p13.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
84464
Gene Symbol
SLX4
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
Location
Chr.16:3581181-3611598 on build GRCh38
Assay Gene Location
Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SLX4 NM_032444.2 12 3644 NP_115820.2
XM_011522715.2 12 3658 XP_011521017.1
XM_017023775.1 9 3049 XP_016879264.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833128 Chr16:3525122 - 3690497 on Build GRCh38 Loss SLX4 NLRC3 TRAP1 LOC101929732 DNASE1 CLUAP1
nsv518509 Chr16:3583656 - 3688960 on Build GRCh38 Gain SLX4 TRAP1 DNASE1
nsv517451 Chr16:3549655 - 3611814 on Build GRCh38 Loss SLX4 NLRC3 LOC101929732 DNASE1

View Full Product Details