Product Details
- Assay Reference Genome
Location
Chr.X:9764614 on build GRCh38
- Cytoband
- Xp22.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs137871560] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 4935
- Gene Symbol
- GPR143
- Gene Name
- G protein-coupled receptor 143
- Gene Aliases
- NYS6, OA1
- Location
- Chr.X:9725413-9765965 on build GRCh38
- Assay Gene Location
- Within Intron 1
View Full Product Details