Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167991.1 | NP_001161463.1 | 4 | 354 | |
NM_133255.2 | NP_573518.2 | 4 | 354 | ||
GenBank mRNA | AK030599.1 | - | 4 | 347 | |
AK042048.1 | - | 4 | 342 | ||
AK136069.1 | - | 4 | 348 | ||
AK145768.1 | - | 4 | 322 | ||
AK161612.1 | - | 4 | 307 | ||
AK166954.1 | - | 4 | 334 | ||
AK169073.1 | - | 4 | 322 | ||
BC019486.1 | - | 4 | 330 | 4211990 | |
BC029967.1 | - | 4 | 295 | ||
BC055881.1 | - | 4 | 300 | 3590336 | |
BC080744.1 | - | 4 | 344 | 30471369 |