Gene Transcripts

Gene Symbol
Mmp24
Entrez Gene ID
17391
Gene Name
matrix metallopeptidase 24
Gene Aliases
AU040325, MT5-MMP, MT5MMP, MTMMP5
Chromosome Location
Chr. 2: 155775344 - 155818366 on Build GRCm38
UniGene ID
Mm.330707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010808.3 NP_034938.3 3, 4 558
GenBank mRNA AB021226.1 - 3, 4 503
AJ010262.1 - 3, 4 504
AY622976.1 - 2, 3 147

View Full Product Details