Gene Transcripts

Gene Symbol
Ccdc73
Entrez Gene ID
211936
Gene Name
coiled-coil domain containing 73
Gene Aliases
2210415I11Rik, 4931417A20, AV080665, BC095976
Chromosome Location
Chr. 2: 104886324 - 104999737 on Build GRCm38
UniGene ID
Mm.416538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177600.4 NP_808268.2 18 3192
GenBank mRNA AK029862.1 - 10 2399
BC150667.1 - 9 2318 9087578

View Full Product Details