Gene Transcripts

Gene Symbol
Ubxn10
Entrez Gene ID
212190
Gene Name
UBX domain protein 10
Gene Aliases
5730509E04Rik, A830047D02, Ubxd3
Chromosome Location
Chr. 4: 138709837 - 138738438 on Build GRCm38
UniGene ID
Mm.292226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285928.1 NP_001272857.1 4 503
NM_001285929.1 NP_001272858.1 3 392
NM_001285930.1 NP_001272859.1 3 256
NM_178671.5 NP_848786.1 2 125
GenBank mRNA AK045964.1 - 2 83
AK046949.1 - 4 504
AK077656.1 - 2 127
AK080662.1 - 2 91
AK133185.1 - 2 111
AK161245.1 - 2 112
AK161296.1 - 2 88
BC027099.1 - 3 256 5041000

View Full Product Details