Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172154.4 | NP_742166.1 | 3 | 3599 | |
GenBank mRNA | AB076079.1 | - | 8 | 1210 | |
AK041090.1 | - | 3 | 3600 | ||
AK041621.1 | - | 8 | 1413 | ||
AK129442.1 | - | 6 | 1035 | ||
AK155007.1 | - | 8 | 1289 | ||
BC050068.1 | - | 3 | 581 | ||
BC090652.1 | - | 5 | 993 | 30657664 | |
BC099938.1 | - | 8 | 1281 | 30085146 |