Gene Transcripts

Gene Symbol
Cog7
Entrez Gene ID
233824
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
5630400E24Rik, Gm167
Chromosome Location
Chr. 7: 121922836 - 121981706 on Build GRCm38
UniGene ID
Mm.442015
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033318.3 NP_001028490.2 1 40
GenBank mRNA AK145163.1 - 1 38
AK154693.1 - 1 42
AK163143.1 - 60
AK172200.1 - 1 42

View Full Product Details