Gene Transcripts

Gene Symbol
Kif6
Entrez Gene ID
319991
Gene Name
kinesin family member 6
Gene Aliases
D130004B10Rik, D130084M03Rik
Chromosome Location
Chr. 17: 49615104 - 49909847 on Build GRCm38
UniGene ID
Mm.441340
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177052.3 NP_796026.2 5 493
GenBank mRNA AB001434.1 - 2 152
AK043404.1 - 5 450
AK083760.1 - 5 530

View Full Product Details