Gene Transcripts

Gene Symbol
Tax1bp1
Entrez Gene ID
52440
Gene Name
Tax1 (human T cell leukemia virus type I) binding protein 1
Gene Aliases
1200003J11Rik, 1700069J21Rik, AA930106, D6Ertd404e, D6Ertd772e, T6bp, TXBP151
Chromosome Location
Chr. 6: 52713575 - 52766780 on Build GRCm38
UniGene ID
Mm.389757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025816.3 NP_080092.2 4 417
GenBank mRNA AK004574.1 - 4 399
AK146076.1 - 4 397
AK171136.1 - 4 384
BC014798.1 - 4 338 3962810

View Full Product Details