Gene Transcripts

Gene Symbol
Lypd6b
Entrez Gene ID
71897
Gene Name
LY6/PLAUR domain containing 6B
Gene Aliases
2310010M24Rik, AW049525
Chromosome Location
Chr. 2: 49787353 - 49948849 on Build GRCm38
UniGene ID
Mm.41381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027990.3 NP_082266.1 10 884
GenBank mRNA AK009282.1 - 10 884
BC126943.1 - 7 628 40090023

View Full Product Details