Gene Transcripts
- Gene Symbol
- Nkd2
- Entrez Gene ID
- 72293
- Gene Name
- naked cuticle 2 homolog (Drosophila)
- Gene Aliases
- 2210403L10Rik, AW212591
- Chromosome Location
- Chr. 13: 73818529 - 73847631 on Build GRCm38
- UniGene ID
- Mm.45506
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028186.4 |
NP_082462.3
|
3, 4 |
378 |
|
GenBank mRNA |
AF358136.1 |
- |
3, 4 |
328 |
|
AK008814.1 |
- |
3, 4 |
360 |
|
AK081904.1 |
- |
2, 3 |
324 |
|
AK145155.1 |
- |
3, 4 |
360 |
|
AK146667.1 |
- |
3, 4 |
360 |
|
AK158350.1 |
- |
3, 4 |
360 |
|
AK165116.1 |
- |
2, 3 |
324 |
|
AK199881.1 |
- |
1, 2 |
40 |
|
AK203955.1 |
- |
1, 2 |
41 |
|
BC019952.1 |
- |
3, 4 |
347 |
4021772 |
View Full Product Details