Gene Transcripts
- Gene Symbol
- Eml4
- Entrez Gene ID
- 78798
- Gene Name
- echinoderm microtubule associated protein like 4
- Gene Aliases
- 4930443C24Rik, AI644019
- Chromosome Location
- Chr. 17: 83350931 - 83480361 on Build GRCm38
- UniGene ID
- Mm.295565
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114361.1 |
NP_001107833.1
|
15, 16 |
1968 |
|
NM_001114362.1 |
NP_001107834.1
|
13, 14 |
1761 |
|
NM_001286567.1 |
NP_001273496.1
|
15, 16 |
1871 |
|
NM_199466.3 |
NP_955760.3
|
12, 13 |
1632 |
|
GenBank mRNA |
AK077527.1 |
- |
7, 8 |
795 |
|
AK144604.1 |
- |
15, 16 |
1969 |
|
BC032258.1 |
- |
1, 2 |
116 |
|
BC065178.1 |
- |
15, 16 |
1809 |
6827397 |
BC066099.1 |
- |
12, 13 |
1563 |
30533573 |
BC067011.1 |
- |
12, 13 |
1581 |
6405184 |
BC070427.1 |
- |
13, 14 |
1701 |
6413041 |
BC079619.1 |
- |
12, 13 |
1563 |
30533573 |
DQ230326.1 |
- |
14, 15 |
1651 |
|
View Full Product Details