Gene Transcripts

Gene Symbol
Srxn1
Entrez Gene ID
76650
Gene Name
sulfiredoxin 1 homolog (S. cerevisiae)
Gene Aliases
1700127B04Rik, AI854065, AW488194, Npn3, Srx, Srx1, TX01
Chromosome Location
Chr. 2: 152105524 - 152111376 on Build GRCm38
UniGene ID
Mm.218639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029688.5 NP_083964.2 2 488
GenBank mRNA AK007296.1 - 2 283
AK089976.1 - 2 285
AK159226.1 - 2 283
AK159613.1 - 2 282
AK161066.1 - 2 325
BC011325.1 - 2 302 3154456
BC049957.1 - 2 488 4507800

View Full Product Details