Gene Transcripts
- Gene Symbol
- Synpo
- Entrez Gene ID
- 104027
- Gene Name
- synaptopodin
- Gene Aliases
- 9030217H17Rik, 9130229N11, 9330140I15Rik, AW046661
- Chromosome Location
- Chr. 18: 60593981 - 60660174 on Build GRCm38
- UniGene ID
- Mm.252321
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020250.1 |
- |
3 |
332 |
|
View Full Product Details