Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025631.3 | NP_079907.2 | 15 | 1409 | |
GenBank mRNA | AK009346.1 | - | 15 | 1381 | |
AK009619.1 | - | 15 | 1382 | ||
AK009754.1 | - | 11 | 985 | ||
AK009776.1 | - | 15 | 1367 | ||
AK009794.1 | - | 15 | 1369 | ||
AK009871.1 | - | 15 | 1378 | ||
AK010130.1 | - | 15 | 1364 | ||
AK010154.1 | - | 15 | 1377 | ||
AK010157.1 | - | 15 | 1408 | ||
AK010215.1 | - | 15 | 1378 | ||
AK028158.1 | - | 15 | 1383 | ||
AK075874.1 | - | 15 | 1380 | ||
AK160516.1 | - | 15 | 1380 | ||
BC116681.1 | - | 15 | 1364 | 40106489 |