Gene Transcripts

Gene Symbol
Lrp1
Entrez Gene ID
16971
Gene Name
low density lipoprotein receptor-related protein 1
Gene Aliases
A2mr, AI316852, CD91, Lrp, b2b1554Clo
Chromosome Location
Chr. 10: 127538157 - 127621148 on Build GRCm38
UniGene ID
Mm.271854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008512.2 NP_032538.2 55 9275
GenBank mRNA AF367720.1 - 55 9246
X67469.1 - 55 9255

View Full Product Details