Gene Transcripts

Gene Symbol
Pdk2
Entrez Gene ID
18604
Gene Name
pyruvate dehydrogenase kinase, isoenzyme 2
Gene Aliases
Chromosome Location
Chr. 11: 95026258 - 95041371 on Build GRCm38
UniGene ID
Mm.29768
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133667.2 NP_598428.2 4 519
GenBank mRNA AF267660.1 - 4 480
AK002647.1 - 4 463
BC021764.1 - 4 473 3488813
CX223857.1 - 4 520

View Full Product Details