Gene Transcripts

Gene Symbol
Kcng2
Entrez Gene ID
240444
Gene Name
potassium voltage-gated channel, subfamily G, member 2
Gene Aliases
AA591524
Chromosome Location
Chr. 18: 80294544 - 80365105 on Build GRCm38
UniGene ID
Mm.279568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190373.1 NP_001177302.1 4 1052
GenBank mRNA AK158780.1 - 1 30

View Full Product Details