Gene Transcripts
- Gene Symbol
- Kif6
- Entrez Gene ID
- 319991
- Gene Name
- kinesin family member 6
- Gene Aliases
- D130004B10Rik, D130084M03Rik
- Chromosome Location
- Chr. 17: 49615104 - 49909847 on Build GRCm38
- UniGene ID
- Mm.441340
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177052.3 |
NP_796026.2
|
18, 19 |
2132 |
|
GenBank mRNA |
AK084082.1 |
- |
3, 4 |
1933 |
|
View Full Product Details